Learn More
Thermo Scientific Chemicals 2'-Amino-2'-deoxyguanosine, 98%
1890.00 NOK - 7790.00 NOK
Chemical Identifiers
CAS 60966-26-9
Molecular Formula C10H14N6O4
Molecular Weight (g/mol) 282.26
MDL Number MFCD01723954
InChI Key ROPTVRLUGSPXNH-SJLGBTOWNA-N
PubChem CID 173775
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15436499
View Documents
Thermo Scientific Chemicals
J64650.MD
250 mg
1890.00 NOK
250mg
15426499
View Documents
Thermo Scientific Chemicals
J64650.03
1 g
7790.00 NOK
1g
Description
Description
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Chemical Identifiers
60966-26-9
282.26
ROPTVRLUGSPXNH-SJLGBTOWNA-N
2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H14N6O4
MFCD01723954
173775
N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
Specifications
60966-26-9
C10H14N6O4
2'-amino-2'-deoxyguanosine, guanosine, 2'-amino-2'-deoxy, 2'-deoxy-2'-aminoguanosine, 2ag, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, guanosine,2'-amino-2'-deoxy, 2'-amino-2'-deoxyguanosine,2'-nh2-dg, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one
N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
282.26
282.26
Powder
White
MFCD01723954
ROPTVRLUGSPXNH-SJLGBTOWNA-N
2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
173775
98%
2'-Amino-2'-deoxyguanosine
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'rtecsNumber' : MF8752000
missing translation for 'tsca' : No
Recommended Storage : Keep cold