Learn More
2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
1765.00 NOK - 6275.00 NOK
Chemical Identifiers
CAS | 2769-71-3 |
---|---|
Molecular Formula | C9H9N |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00013479 |
InChI Key | DNJLFZHMJDSJFN-UHFFFAOYSA-N |
Synonym | 2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanideShow More |
PubChem CID | 76009 |
IUPAC Name | 2-isocyano-1,3-dimethylbenzene |
SMILES | CC1=CC=CC(C)=C1[N+]#[C-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337967
|
Thermo Scientific Alfa Aesar
020748.03 |
1 g |
1765.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347967
|
Thermo Scientific Alfa Aesar
020748.06 |
5 g |
6275.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-Dimethylphenyl isocyanate is used in the preparation of homoleptic metallaamidine complex, derivatized bets-cyclodextrins. It is used as chiral stationary phase in normal-phase liquid chromatography. It is also involved in the preparation of tris( 2,6-dimethylphenylimido)methylrhenium (VII) and 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2769-71-3 | |
131.18 | |
DNJLFZHMJDSJFN-UHFFFAOYSA-N | |
76009 | |
CC1=CC=CC(C)=C1[N+]#[C-] |
C9H9N | |
MFCD00013479 | |
2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanideShow More | |
2-isocyano-1,3-dimethylbenzene |
Specifications
73°C to 74°C | |
Unpleasant | |
C9H9N | |
UN2811 | |
Insoluble in water. | |
CC1=CC=CC(C)=C1[N+]#[C-] | |
131.18 | |
131.18 |
2769-71-3 | |
Crystalline | |
MFCD00013479 | |
2,6-dimethylphenyl isocyanide, 2,6-dimethylphenylisonitrile, 2,6-dimethylphenylisocyanide, 2,6-xyleneisonitrile, 2,6-xylyl isocyanide, benzene,2-isocyano-1,3-dimethyl, benzene, 2-isocyano-1,3-dimethyl-9ci, vic.-m-xylyl isocyanide, acmc-20ao80, 2,6-dimethyiphenyl isocyanide | |
DNJLFZHMJDSJFN-UHFFFAOYSA-N | |
2-isocyano-1,3-dimethylbenzene | |
76009 | |
2,6-Dimethylphenyl isocyanide |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
H301+H311-H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 220-459-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 2,6-Dimethylphenyl isocyanide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H330-Fatal if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 2,6-Xylyl isocyanide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.