missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzonitrile, 99%, Thermo Scientific™

781.00 NOK - 3401.00 NOK

Chemical Identifiers

CAS 6575-13-9
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
InChI Key QSACPWSIIRFHHR-UHFFFAOYSA-N
Synonym benzonitrile, 2,6-dimethyl, unii-1zh37s5b0z, 2,6-dimethylbenzenecarbonitrile, 2,6-dimethyl-benzonitrile, 2-cyano-m-xylene, ksc494c5t, 2,6-dimethylbenz0nitrile, 3-cyano-2,4-dimethylphenylShow MoreShow Less
PubChem CID 81057
IUPAC Name 2,6-dimethylbenzonitrile
SMILES CC1=C(C(=CC=C1)C)C#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15575076
View Documents
Thermo Scientific Acros
355590050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15325736
View Documents
Thermo Scientific Acros
355590250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dimethylbenzonitrile, 99%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.