missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11411818
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-07-1
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002462
InChI Key AKEUNCKRJATALU-UHFFFAOYSA-N
Synonym gamma-resorcylic acid, 2-carboxyresorcinol, 2,6-resorcylic acid, 6-hydroxysalicylic acid, benzoic acid, 2,6-dihydroxy, .gamma.-resorcylic acid, unii-rsa5g6vrpv, rsa5g6vrpv, 2,6-dihydroxy benzoic acid, 2,6-dihydroxy-benzoic acid
PubChem CID 9338
ChEBI CHEBI:68465
IUPAC Name 2,6-dihydroxybenzoic acid
SMILES OC(=O)C1=C(O)C=CC=C1O

Specifications

Melting Point ∼164°C (decomposition)
Quantity 500 g
Beilstein 2209755
Formula Weight 154.12
Percent Purity 98%
Chemical Name or Material 2,6-Dihydroxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'