Learn More
2,6,8-Trimethyl-4-nonanol, erythro + threo, 90+%, Thermo Scientific Chemicals
784.00 NOK - 1992.00 NOK
Chemical Identifiers
CAS | 123-17-1 |
---|---|
Molecular Formula | C12H26O |
Molecular Weight (g/mol) | 186.339 |
MDL Number | MFCD00026491 |
InChI Key | LFEHSRSSAGQWNI-UHFFFAOYSA-N |
Synonym | 2,6,8-trimethyl-4-nonanol, 4-nonanol, 2,6,8-trimethyl, 4-hydroxy-2,6,8-trimethylnonane, 2,6,8-trimethylnonanol-4, 2,4,8-trimethyl-6-nonanol, dsstox_cid_9159, dsstox_rid_78690, dsstox_gsid_29159, acmc-1bphg, 2,8-trimethylnonanol-4 |
PubChem CID | 61056 |
IUPAC Name | 2,6,8-trimethylnonan-4-ol |
SMILES | CC(C)CC(C)CC(CC(C)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11345329
|
Thermo Scientific Alfa Aesar
B20830.06 |
5 g |
784.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355329
|
Thermo Scientific Alfa Aesar
B20830.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-17-1 | |
186.339 | |
LFEHSRSSAGQWNI-UHFFFAOYSA-N | |
61056 | |
CC(C)CC(C)CC(CC(C)C)O |
C12H26O | |
MFCD00026491 | |
2,6,8-trimethyl-4-nonanol, 4-nonanol, 2,6,8-trimethyl, 4-hydroxy-2,6,8-trimethylnonane, 2,6,8-trimethylnonanol-4, 2,4,8-trimethyl-6-nonanol, dsstox_cid_9159, dsstox_rid_78690, dsstox_gsid_29159, acmc-1bphg, 2,8-trimethylnonanol-4 | |
2,6,8-trimethylnonan-4-ol |
Specifications
123-17-1 | |
C12H26O | |
MFCD00026491 | |
LFEHSRSSAGQWNI-UHFFFAOYSA-N | |
2,6,8-trimethylnonan-4-ol | |
61056 | |
≥90% |
123°C (20 mmHg) | |
1.435 | |
2,6,8-trimethyl-4-nonanol, 4-nonanol, 2,6,8-trimethyl, 4-hydroxy-2,6,8-trimethylnonane, 2,6,8-trimethylnonanol-4, 2,4,8-trimethyl-6-nonanol, dsstox_cid_9159, dsstox_rid_78690, dsstox_gsid_29159, acmc-1bphg, 2,8-trimethylnonanol-4 | |
CC(C)CC(C)CC(CC(C)C)O | |
186.339 | |
184.32 | |
2,6,8-Trimethyl-4-nonanol, erythro + threo |
Safety and Handling
missing translation for 'einecsNumber' : 204-606-8
missing translation for 'rtecsNumber' : RA7950000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.