Learn More
Thermo Scientific Alfa Aesar 2,5LT n-Hexane, HPLC Grade, 95% min 2.5l
Brand: Thermo Scientific Alfa Aesar 039199.K4
2,5LT n-Hexane, HPLC Grade, 95% min 2.5l
Description
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2529297 | Certificate of Analysis | 05/03/2025 | 039199.K4,039199.0F |
X11K943 | Certificate of Analysis | 11/11/2024 | 039199.K7,039199.K4,039199.K2,039199.0F,039199.M1 |
X30K983 | Certificate of Analysis | 11/11/2024 | 039199.K7,039199.K4,039199.K2,039199.0F,039199.M1 |
X11K947 | Certificate of Analysis | 18/10/2024 | 039199.K7,039199.K4,039199.K2,039199.0F,039199.M1 |
W30K945 | Certificate of Analysis | 03/10/2024 | 039199.K7,039199.K4,039199.K2,039199.0F,039199.M1 |