Learn More
2,5-Dihydroxy-1,4-dithiane, 96% Product Code.: 11447566
2,5-Dihydroxy-1,4-dithiane, 96%
Product Code. 11447566
1000 g, 1000g
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
Product Code. 11447566
Brand: Thermo Scientific Alfa Aesar A14620.0B
Request Bulk or Custom Format
CAS: 40018-26-6 | C4H8O2S2 | 152.226 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40018-26-6
Molecular Formula C4H8O2S2
Molecular Weight (g/mol) 152.226
MDL Number MFCD00006659
InChI Key YUIOPHXTILULQC-UHFFFAOYSA-N
Synonym p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme
PubChem CID 98330
IUPAC Name 1,4-dithiane-2,5-diol
SMILES C1C(SCC(S1)O)O
Specifications
Melting Point ∼130°C (decomposition)
Quantity 1000 g
UN Number UN2811
Beilstein 104402
Formula Weight 152.24
Percent Purity 96%
Chemical Name or Material 2,5-Dihydroxy-1,4-dithiane
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number10251131 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeA14620.30,A14620.0B,A14620.18
Lot Number10219851 Certificate TypeCertificate of Analysis Date29/11/2022 Product CodeA14620.30,A14620.0B,A14620.18
Lot Number10223749 Certificate TypeCertificate of Analysis Date21/06/2022 Product CodeA14620.30,A14620.0B,A14620.18
Lot Number10122184 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA14620.30,A14620.0B,A14620.18
Lot Number10202200 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA14620.30,A14620.0B,A14620.18
Product Title
2,5-Dihydroxy-1,4-dithiane, 96% >