missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Toluoyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 11445153
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-55-2
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.26
MDL Number MFCD00020287
InChI Key ICQOWIXIHDDXDI-UHFFFAOYSA-N
Synonym 2-4-methylbenzoyl benzoic acid, 2-p-toluoyl benzoic acid, benzoic acid, 2-4-methylbenzoyl, 2-p-toluyl benzoic acid, o-p-toluoyl benzoic acid, benzoic acid, o-p-toluoyl, 2-p-toluoylbenzoic acid, unii-4dn9g70pqf, 4'-methylbenzophenone-2-carboxylic acid, p-toluoyl-o-benzoic acid
PubChem CID 66563
IUPAC Name 2-(4-methylbenzoyl)benzoic acid
SMILES CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1C(O)=O

Specifications

Melting Point 137°C to 139°C
Quantity 100 g
Beilstein 2111078
Merck Index 14,9539
Formula Weight 240.26
Percent Purity 98%
Chemical Name or Material 2-(4-Toluoyl)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.