missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dimethoxybenzylamine, 98% Product Code.: 10558343

2,4-Dimethoxybenzylamine, 98%

Product Code. p-3211523 Shop All Thermo Scientific Chemicals Products
5 mL, Glass bottle, 5mL
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10558343

Brand: Thermo Scientific Acros 369950050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20781-20-8 | C9H13NO2 | 167.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20781-20-8
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00052393
InChI Key QOWBXWFYRXSBAS-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine
PubChem CID 597250
IUPAC Name (2,4-dimethoxyphenyl)methanamine
SMILES COC1=CC(=C(C=C1)CN)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1130g/mL
Boiling Point 140°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2NH2
Refractive Index 1.5480 to 1.55
Quantity 5 mL
Specific Gravity 1.113
Formula Weight 167.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dimethoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dimethoxybenzylamine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.