missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dimethoxyaniline, 97%, Thermo Scientific™ Product Code.: 11431678

2,4-Dimethoxyaniline, 97%, Thermo Scientific™

Product Code. 11431678
50 g, 50g
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11431678

Brand: Thermo Scientific Alfa Aesar A13943.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Various 2,4-dimethoxyphenylsemicarbazones were synthesized starting from 2,4-dimethoxyaniline via a phenylcarbamate intermediate. The hydrochloride of 2,4-dimethoxyaniline was prepared by intro- ducing hydrogen chloride gas into an etheral solution and washing the precipitate with absolute diethyl ether.

Solubility
Slightly soluble in water(7g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. May be incompatible with isocyanates, halogenated organics, peroxides, phenols (acidic), epoxides, anhydrides, and acid halides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2735-04-8
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008371
InChI Key GEQNZVKIDIPGCO-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dimethoxy, aniline, 2,4-dimethoxy, 2,4-dimethoxyphenylamine, unii-i079354gsi, benzenamine, 2,4-dimethoxy-, hydrochloride, 2,4-domethoxyaniline, 2,4-dimethoxy-anilin, 2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline
PubChem CID 17652
IUPAC Name 2,4-dimethoxyaniline
SMILES COC1=CC(=C(C=C1)N)OC

Specifications

Melting Point 32°C to 37°C
Density 1.075
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 50 g
UN Number UN2811
Beilstein 638704
Sensitivity Light sensitive
Solubility Information Slightly soluble in water(7g/L).
Formula Weight 153.18
Percent Purity 97%
Chemical Name or Material 2,4-Dimethoxyaniline
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dimethoxyaniline, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.