Learn More
2,4-Dichlorobenzenesulfonyl chloride, 98%, Thermo Scientific™
955.00 NOK - 7223.00 NOK
Chemical Identifiers
CAS | 16271-33-3 |
---|---|
Molecular Formula | C6H3Cl3O2S |
Molecular Weight (g/mol) | 245.498 |
MDL Number | MFCD00052712 |
InChI Key | FDTPBIKNYWQLAE-UHFFFAOYSA-N |
Synonym | 2,4-dichloro-benzenesulfonyl chloride, 2,4-dichlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dichloro, 2,4-dichlorobenzenesulphonyl chloride, 2,4-dichlorobenzenesulfonylchloride, 2,4-dichlorophenylsulfonyl chloride, 2,4-dichlorobenzenesulphonylchloride, 2,4-dichlorophenyl chlorosulfone, pubchem5115, acmc-1c9b8 |
PubChem CID | 204419 |
IUPAC Name | 2,4-dichlorobenzenesulfonyl chloride |
SMILES | C1=CC(=C(C=C1Cl)Cl)S(=O)(=O)Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
16271-33-3 | |
245.498 | |
FDTPBIKNYWQLAE-UHFFFAOYSA-N | |
204419 | |
C1=CC(=C(C=C1Cl)Cl)S(=O)(=O)Cl |
C6H3Cl3O2S | |
MFCD00052712 | |
2,4-dichloro-benzenesulfonyl chloride, 2,4-dichlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dichloro, 2,4-dichlorobenzenesulphonyl chloride, 2,4-dichlorobenzenesulfonylchloride, 2,4-dichlorophenylsulfonyl chloride, 2,4-dichlorobenzenesulphonylchloride, 2,4-dichlorophenyl chlorosulfone, pubchem5115, acmc-1c9b8 | |
2,4-dichlorobenzenesulfonyl chloride |
Specifications
16271-33-3 | |
C6H3Cl3O2S | |
UN3261 | |
Moisture sensitive | |
FDTPBIKNYWQLAE-UHFFFAOYSA-N | |
2,4-dichlorobenzenesulfonyl chloride | |
204419 | |
98% |
50°C to 58°C | |
MFCD00052712 | |
2806192 | |
2,4-dichloro-benzenesulfonyl chloride, 2,4-dichlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dichloro, 2,4-dichlorobenzenesulphonyl chloride, 2,4-dichlorobenzenesulfonylchloride, 2,4-dichlorophenylsulfonyl chloride, 2,4-dichlorobenzenesulphonylchloride, 2,4-dichlorophenyl chlorosulfone, pubchem5115, acmc-1c9b8 | |
C1=CC(=C(C=C1Cl)Cl)S(=O)(=O)Cl | |
245.498 | |
245.51 | |
2,4-Dichlorobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : DB8927500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.