missing translation for 'onlineSavingsMsg'
Learn More
2,4,6-Triiodophenol, 98%, Thermo Scientific Chemicals Product Code.: 11423367

2,4,6-Triiodophenol, 98%, Thermo Scientific Chemicals

Product Code. 11423367
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11423367

Brand: Thermo Scientific Alfa Aesar A17145.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 609-23-4 | C6H3I3O | 471.802 g/mol

2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.

Solubility
Insoluble in water.

Notes
Incompatible withacid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-23-4
Molecular Formula C6H3I3O
Molecular Weight (g/mol) 471.802
MDL Number MFCD00002179
InChI Key VAPDZNUFNKUROY-UHFFFAOYSA-N
Synonym phenol, 2,4,6-triiodo, triiodophenol, unii-9rb2r81a7u, 2,4,6-trijodfenol, 2,4,6-trijodfenol czech, 2,4,6-triiodo-phenol, chembl81721, 2,4,6-triiodo-phenol tip, 2,6-triiodophenol, phenol,4,6-triiodo
PubChem CID 11862
IUPAC Name 2,4,6-triiodophenol
SMILES C1=C(C=C(C(=C1I)O)I)I

Specifications

Melting Point 156°C to 160°C
Quantity 25 g
Beilstein 2046861
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 471.8
Percent Purity 98%
Chemical Name or Material 2,4,6-Triiodophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4,6-Triiodophenol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.