missing translation for 'onlineSavingsMsg'
Learn More
2-(3-Methoxyallylidene)malonic acid dimethyl ester, 95% Product Code.: 10490911

2-(3-Methoxyallylidene)malonic acid dimethyl ester, 95%

Product Code. p-4510508 Shop All Thermo Scientific Chemicals Products
2.5 g, Glass bottle, 2.50g
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.50g
This item is not returnable. View return policy

Product Code. 10490911

Brand: Thermo Scientific Acros 302090025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41530-32-9 | C9H12O5 | 200.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41530-32-9
Molecular Formula C9H12O5
Molecular Weight (g/mol) 200.19
InChI Key NOBNTZJTOBIOHL-GQCTYLIASA-N
Synonym 2-3-methoxyallylidene malonic acid dimethyl ester, dimethyl 2-3-methoxyallylidene malonate, 1,3-dimethyl 2-2e-3-methoxyprop-2-en-1-ylidene propanedioate, 2-3-methoxyallylidene malonic acid dimethyl ester, 96, dimethyl 2-e-3-methoxyprop-2-enylidene propanedioate, propanedioic acid, 3-methoxy-2-propenylidene-, dimethyl ester, dimethyl e-2-3-methoxyallylidene malonate, 3-methoxy-2-propenylidene malonic acid dimethyl ester, dimethyl 2-2e-3-methoxy-2-propenylidene malonate #
PubChem CID 5374172
IUPAC Name dimethyl 2-[(E)-3-methoxyprop-2-enylidene]propanedioate
SMILES COC=CC=C(C(=O)OC)C(=O)OC

Specifications

Color Green-Yellow
Boiling Point 120.0°C to 125.0°C (0.3 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5370 to 1.5390
Quantity 2.5 g
Formula Weight 200.19
Percent Purity 95%
Physical Form Low Melting Solid
Chemical Name or Material 2-(3-Methoxyallylidene)malonic acid dimethyl ester
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0394753 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code302090025,302090100
Lot NumberA0394753 Certificate TypeCertificate of Analysis Date12/09/2022 Product Code302090025,302090100

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(3-Methoxyallylidene)malonic acid dimethyl ester, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.