missing translation for 'onlineSavingsMsg'
Learn More
2,3-Difluorocinnamic acid, 98%, Thermo Scientific™ Product Code.: 11440621

2,3-Difluorocinnamic acid, 98%, Thermo Scientific™

Product Code. 11440621
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11440621

Brand: Thermo Scientific Alfa Aesar B20105.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207981-48-4
Molecular Formula C9H6F2O2
Molecular Weight (g/mol) 184.142
MDL Number MFCD00061291
InChI Key NVQHKPLMLCHFSU-SNAWJCMRSA-N
Synonym 2,3-difluorocinnamic acid, trans-2,3-difluorocinnamic acid, e-3-2,3-difluorophenyl acrylic acid, 3-2,3-difluorophenyl acrylic acid, 2e-3-2,3-difluorophenyl prop-2-enoic acid, e-3-2,3-difluorophenyl prop-2-enoic acid, 3-2,3-difluoro-phenyl-acrylic acid, 2e-3-2,3-difluorophenyl-2-propenoic acid, pubchem4329, rarechem bk hw 0120
PubChem CID 5702854
IUPAC Name (E)-3-(2,3-difluorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C(=C1)F)F)C=CC(=O)O

Specifications

Melting Point 198°C to 201°C
Quantity 1 g
Formula Weight 184.14
Percent Purity 98%
Chemical Name or Material 2,3-Difluorocinnamic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10145607 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10165967 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Difluorocinnamic acid, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.