missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-6-methoxybenzylamine, 97%, Thermo Scientific™

Product Code. 15463517
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886501-77-5
Molecular Formula C8H9F2NO
Molecular Weight (g/mol) 173.163
MDL Number MFCD04116022
InChI Key TYGWYXQDCGKHHY-UHFFFAOYSA-N
Synonym 2,3-difluoro-6-methoxybenzylamine, 2,3-difluoro-6-methoxyphenyl methanamine, 1-2,3-difluoro-6-methoxyphenyl methanamine, 2,3-difluoro-6-methoxyphenyl methylamine
PubChem CID 4339538
IUPAC Name (2,3-difluoro-6-methoxyphenyl)methanamine
SMILES COC1=C(C(=C(C=C1)F)F)CN

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 173.16
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-6-methoxybenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'