missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-6-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

1278.00 NOK - 4219.00 NOK

Chemical Identifiers

CAS 886501-99-1
Molecular Formula C8H4BrCl2F3
Molecular Weight (g/mol) 307.919
MDL Number MFCD06660275
InChI Key BTXOBZONYMZJCP-UHFFFAOYSA-N
Synonym 2,3-dichloro-6-trifluoromethyl benzyl bromide, 2-bromomethyl-3,4-dichloro-1-trifluoromethyl benzene
PubChem CID 17750717
IUPAC Name 3-(bromomethyl)-1,2-dichloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C(=C1C(F)(F)F)CBr)Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387029
SDS View Documents
Thermo Scientific Alfa Aesar
H26859.03
1 g N/A N/A N/A
11397029
SDS View Documents
Thermo Scientific Alfa Aesar
H26859.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Dichloro-6-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.