missing translation for 'onlineSavingsMsg'
Learn More
2,3,4-Trifluorobenzoyl chloride, 97%, Thermo Scientific™ Product Code.: 11415487

2,3,4-Trifluorobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11415487
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11415487

Brand: Thermo Scientific Alfa Aesar B23484.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157373-08-5
Molecular Formula C7H2ClF3O
Molecular Weight (g/mol) 194.537
MDL Number MFCD00075078
InChI Key NXRQXCFBZGIRGN-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzoylchloride, 2,3,4-trifluoro-benzoyl chloride, benzoyl chloride, 2,3,4-trifluoro, benzoyl chloride, 2,3,4-trifluoro-9ci, trifluorobenzoylchloride, trifluorobenzoyl chloride, intermediates-zcf02190, acmc-209dg2, 2,3,4-trifluorobenzoyl chlorideShow MoreShow Less
PubChem CID 2734022
IUPAC Name 2,3,4-trifluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1C(=O)Cl)F)F)F

Specifications

Density 1.522
Boiling Point 143°C to 144°C (130 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.497
Quantity 1 g
UN Number UN3265
Beilstein 9122972
Sensitivity Moisture sensitive
Formula Weight 194.54
Percent Purity 97%
Chemical Name or Material 2,3,4-Trifluorobenzoyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10144297 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10182798 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10044333 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3,4-Trifluorobenzoyl chloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.