missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluorobenzoyl chloride, 98%, Thermo Scientific™

Product Code. 11490498
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94695-48-4
Molecular Formula C7HClF4O
Molecular Weight (g/mol) 212.53
MDL Number MFCD00075164
InChI Key XWCKIXLTBNGIHV-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzoylchloride, tetrafluorobenzoyl chloride, 2,3,4,5-tetrafluoro benzoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobezoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, 2,3,4,5-tetrafluorobezoylchloride
PubChem CID 2733689
IUPAC Name 2,3,4,5-tetrafluorobenzoyl chloride
SMILES FC1=CC(C(Cl)=O)=C(F)C(F)=C1F

Specifications

Density 1.58
Boiling Point 65°C to 66°C (10 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.4787
Quantity 1 g
UN Number UN3265
Beilstein 5266860
Sensitivity Moisture sensitive
Formula Weight 212.53
Percent Purity 98%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzoyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3,4,5-Tetrafluorobenzoyl chloride, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.