missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Naphthoyl)-1-(m-toluoyl)hydrazine, 98%, Thermo Scientific™

Product Code. 11365219
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83803-96-7
Molecular Formula C19H16N2O2
Molecular Weight (g/mol) 304.349
MDL Number MFCD00021628
InChI Key NOZDWMYGSUBAEE-UHFFFAOYSA-N
Synonym 2-2-naphthoyl-1-m-toluoyl hydrazine, 2'-3-methylbenzoyl-2-naphthohydrazide, n'-3-methylbenzoyl naphthalene-2-carbohydrazide, n'-3-methylbenzoyl-2-naphthohydrazide, n'-3-methylbenzoyl-2-naphthalenecarbohydrazide, 3-methyl-n'-naphthalene-2-carbonyl benzohydrazide, 1-3-methylbenzoyl-2-2-naphthoyl hydrazine, n-3-methylphenyl carbonylamino-2-naphthylcarboxamide, 2-naphthalenecarboxylic acid, n'-3-methylbenzoyl hydrazide, 2-naphthalenecarboxylicacid, 2-3-methylbenzoyl hydrazide
PubChem CID 96763
IUPAC Name N'-(3-methylbenzoyl)naphthalene-2-carbohydrazide
SMILES CC1=CC=CC(=C1)C(=O)NNC(=O)C2=CC3=CC=CC=C3C=C2

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 304.33
Percent Purity 98%
Chemical Name or Material 2-(2-Naphthoyl)-1-(m-toluoyl)hydrazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(2-Naphthoyl)-1-(m-toluoyl)hydrazine, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.