missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-amine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | 2,2-dimethyl-3H-1-benzofuran-7-amine |
Formula Weight | 163.22 |
Percent Purity | 95% |
Chemical Name or Material | 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-amine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-amine, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction