missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate, 98%, Thermo Scientific Chemicals
398.57 NOK - 1998.73 NOK
Chemical Identifiers
CAS | 1517-82-4 |
---|---|
Molecular Formula | C17H26O2S |
Molecular Weight (g/mol) | 294.46 |
InChI Key | NQICGNSARVCSGJ-DSRNLFJRSA-N |
Synonym | 1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester |
PubChem CID | 101348873 |
IUPAC Name | [(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate |
SMILES | CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11915751
|
Thermo Scientific Acros
428110010 |
1 g | Glass bottle |
398.57 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11925751
|
Thermo Scientific Acros
428110100 |
10 g | Glass bottle |
1998.73 NOK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1517-82-4 | |
294.46 | |
1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl esterShow More | |
[(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate |
C17H26O2S | |
NQICGNSARVCSGJ-DSRNLFJRSA-N | |
101348873 | |
CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C |
Specifications
1517-82-4 | |
Brown to Yellow or Gray | |
97.5% min. (HPLC) | |
−190° to −204° (20°C, 589nm) (c=2, acetone) | |
NQICGNSARVCSGJ-DSRNLFJRSA-N | |
[(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate | |
101348873 | |
98% | |
(1R, 2S, 5R)-(-)-Menthyl (S)-p-toluenesulfinate |
102.0°C to 104.0°C | |
Authentic | |
C17H26O2S | |
1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester | |
CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C | |
294.46 | |
294.46 | |
Solution |
Safety and Handling
Product Identifier
- (1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate, 98%, Thermo Scientific Chemicals