missing translation for 'onlineSavingsMsg'
Learn More
1H,1H,2H-Perfluoro-1-decene, 99%, Thermo Scientific™ Product Code.: 11345299

1H,1H,2H-Perfluoro-1-decene, 99%, Thermo Scientific™

Product Code. p-7024956
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11345299

Brand: Thermo Scientific Alfa Aesar B20231.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H-Perfluoro-1-decene can be used to prepare perfluorononan-1-oic acid. Appliance Paint, Boat Paint, Buidling. It con be used as monomer and key intermediate of fluoro silicon surfactant.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21652-58-4
Molecular Formula C10H3F17
Molecular Weight (g/mol) 446.11
MDL Number MFCD00039246
InChI Key NKAMGQZDVMQEJL-UHFFFAOYSA-N
Synonym 1h,1h,2h-perfluoro-1-decene, perfluoro-n-octyl ethylene, perfluorooctyl ethylene, tridecafluoro 1-decene, 1h,1h,2h-perfluorodec-1-ene, 1h,1h,2h-heptadecafluoro-1-decene, perfluoroctyl ethylene, cf3 cf2 7ch=ch2, 1-heptadecafluorooctyl ethene
PubChem CID 88990
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C

Specifications

Density 1.68
Boiling Point 146°C to 147°C
Flash Point 125°C (257°F)
Linear Formula CF3(CF2)7CH=CH2
Refractive Index 1.3033
Quantity 5 g
Beilstein 2066558
Solubility Information Insoluble in water.
Formula Weight 446.11
Percent Purity 99%
Chemical Name or Material 1H,1H,2H-Perfluoro-1-decene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10126060 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number90005901 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number90008313 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number90008391 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H,1H,2H-Perfluoro-1-decene, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.