Learn More
Bochem™ 18/10 Stainless Steel Support Rod
Manufactured from 18/10 stainless steel. BOCHEM™ 18/10 Stainless Steel Support Rod come with M10 thread.
Brand:  Bochem™ 5113
Product Code. 10529091
  • 332.00 NOK / Each
Only null left
Estimated Shipment: 14-07-2025
Description
  • Stainless steel
  • With thread M10
Specifications
Specifications
Stainless Steel (18/10)
Support Rod
1 Ea.
1000 mm
Stand bases with thread M10
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Steel Support Rod > Diameter: 12mm