Learn More
Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim
Evaporate materials in the laboratory wit the Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim.
Brand: Bochem™ 8459
This item is not returnable.
View return policy
Description
- 18/10 stainless steel
- Substantial rim size
- Rectangular shape
Specifications
Flat | |
Rectangular | |
375 mm |
Stainless Steel | |
Evaporating Dish | |
45 mm |
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim > Width: 270mm