Learn More
Agencinox™ 18/10 Stainless Steel Evaporating Dish
Use these dishes for general evaporation applications. Agencinox™ 18/10 Stainless Steel Evaporating Dishes are available in a variety of sizes.
174.00 NOK - 240.00 NOK
Specifications
Bottom Shape | Flat |
---|---|
Material | Stainless Steel |
Shape | Round |
Type | Evaporating Dish |
For Use With (Application) | Evaporating Application |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11761923
|
Agencinox™
406 |
50 mL |
174.00 NOK
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
11771923
|
Agencinox™
408 |
100 mL |
240.00 NOK
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
Description
- Low form evaporating dish
- Stainless steel construction
Specifications
Flat | |
Round | |
Evaporating Application |
Stainless Steel | |
Evaporating Dish | |
20 mm |
Product Title
Agencinox™ 18/10 Stainless Steel Evaporating Dish