Learn More
Bochem™ 18/10 Stainless Mini Funnels
Brand:  Bochem™ 8855
Product Code. 12651895
  • 51.15 NOK / Each
Only null left
Estimated Shipment: 20-08-2025
Specifications
4/27 mm
Stainless Steel (18/10)
50 mm
Mini Funnel
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Mini Funnels > Diameter: 0.148mm