Learn More
Thermo Scientific Alfa Aesar 1GR 3-(2-Chloroethylcarbamoyl)benzeneboronic acid, 96% 1g
Brand: Thermo Scientific Alfa Aesar H53342.03
1GR 3-(2-Chloroethylcarbamoyl)benzeneboronic acid, 96% 1g
Additional Details : CAS Number : 874288-12-7 Weight : 0.00100kg
Description

Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
B-BD010201 | Certificate of Analysis | 07/12/2021 | |
B-BD010211 | Certificate of Analysis | 07/12/2021 |