Learn More
Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments
Ensure reliable connections for Thermo Scientific™ GC instruments with these Thermo Scientific™ Ferrules.
Brand:  Thermo Scientific™ 290VT187
789.32 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10630826
  • 793.27 NOK / Pack of 10
Estimated Shipment: 11-06-2025
Description
Use Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments to ensure optimal performance of your Thermo Scientific GC instrument. They are available in three different materials and in various dimensions to accommodate a range of instruments and applications. All ferrules are are supplied in contaminant-free, individual blister packs.
Features
Available in 100% graphite, 15% graphite/85% vespel and SilTite metal
Ferrules for different types of injectors and detectors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferrule
0.32 mm
15% Graphite, 85% Vespel
290 VT187
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments > 0.32mm I.D.; For Thermo Trace and Focus Injectors