Learn More
Keystone Cleanroom Products 1422A Tyvek™ Elastic Equipment Cover
BIOPHARMAGREENER_CHOICE
1422A Tyvek™ Elastic Equipment Cover is an ideal choice in pharmaceutical operations to help keep clean equipment protected from both environmental and product cross contamination.
16885.00 NOK - 66055.00 NOK
Specifications
Product Type Cover
Color White
Material Tyvek
View More Specs
Products 24
Product Code Brand Length (English) Width (English) Height (English) Price Quantity & Availability  
Product Code Brand Length (English) Width (English) Height (English) Price Quantity & Availability  
17778267
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-201608E
20 in. 20 in. 8 in.
16885.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17708277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-281406E
28 in. 28 in. 5 in.
16885.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17718277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-282050E
28 in. 28 in. 50 in.
55800.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17728277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-301616E
30 in. 16 in. 16 in.
39655.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17738277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-301840E
30 in. 30 in. 40 in.
52850.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17748277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-302212E
30 in. 22 in. 12 in.
43385.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17708287
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-3260E
32 in. 32 in. 60 in.
48435.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17738287
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-343442E
34 in. 34 in. 42 in.
55060.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17748287
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-362660E
36 in. 26 in. 60 in.
51640.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
18066991
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-423216E
42 in. 32 in. 16 in.
38335.00 NOK
Pack of 50
17778287
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-423460E
42 in. 34 in. 60 in.
47540.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17718297
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-444460E
44 in. 44 in. 60 in.
57270.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17738297
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-482430E
48 in. 24 in. 30 in.
49910.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17728307
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-505014E
50 in. 50 in. 14 in.
45490.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17768307
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-545486E
54 in. 54 in. 86 in.
63685.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17778307
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-563660E
56 in. 36 in. 60 in.
56800.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17788307
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-602854E
60 in. 28 in. 54 in.
56740.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17708317
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-603072E
60 in. 30 in. 72 in.
55480.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17718317
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-604054E
60 in. 40 in. 54 in.
58740.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17738317
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-604444E
60 in. 44 in. 44 in.
52850.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17758317
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-605060E
60 in. 50 in. 60 in.
52850.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17778317
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-607860E
60 in. 78 in. 60 in.
66055.00 NOK
Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17788317
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-642478E
64 in. 24 in. 78 in.
30240.00 NOK
Pack of 25
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17798317
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-742686E
74 in. 26 in. 86 in.
34790.00 NOK
Pack of 25
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
Description
Description
  • Keeps Cleaned Equipment Clean and Protected from Contamination
  • Breathable
  • Form Fitting, Fast and Consistent
  • Manufactured and packaged in a ISO 6 cleanroom
  • Conveniently packaged: 1 heat-sealed labeled poly bag
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Cover
White
Tyvek