Learn More
Fisherbrand™ 13mm PP Screw Cap, 13-425 thread
Polypropylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 13 08 0452
Description
- 13mm
- PP Screw Cap
- Closed top
Specifications
White | |
Polypropylene | |
ND13 |
13 mm | |
Closed Top Screw Cap |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30