missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fesoterodine fumarate, MedChemExpress

Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
Description
Fesoterodine Fumarate is an orally active, nonsubtype selective, competitive muscarinic receptor (mAChR) antagonist with pKi values of 8.0, 7.7, 7.4, 7.3, 7.5 for M1, M2, M3, M4, M5 receptors, respectively. Fesoterodine Fumarate is used for the overactive bladder (OAB).
Specifications
Specifications
Chemical Name or Material | Fesoterodine fumarate |
CAS | 286930-03-8 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C30H41NO7 |
Quantity | 10 mg |
Solubility Information | DMSO : 100 mg/mL (189.52 mM; Need ultrasonic) ∣H2O : 100 mg/mL (189.52 mM; Need ultrasonic) |
SMILES | OCC1=CC([C@H](CCN(C(C)C)C(C)C)C2=CC=CC=C2)=C(C=C1)OC(C(C)C)=O.OC(/C=C/C(O)=O)=O |
Molecular Weight (g/mol) | 527.65 |
Formula Weight | 527.65 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Fesoterodine fumarate, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction