Learn More
Tocris Bioscience™ BIX 02189
Selective MEK5 and ERK5 inhibitor
Brand:  Tocris Bioscience™ 4842/10
 View more versions of this product
Product Code. 18756846
  • 3095.00 NOK / 10mg
Description
BIX 02189 is a selective MEK5 and ERK5 inhibitor (IC50 values are 1.5 and 59 nM respectively). Inhibits sorbitol-induced ERK5 phosphorylation in HeLa cells. Selective for MEK5 and ERK5 over MEK1/2, ERK1/2 and a panel of 87 kinases. Induces apoptosis in acute myeloid leukemia tumor cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
>98%
BIX 02189
BIX 02189
10 mg
440.54
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Tocris Bioscience™ BIX 02189 > 10mg
For Research Use Only