Learn More
Thermo Scientific Alfa Aesar 100GR Silicone oil, for oil baths, usable rangefrom -40 to +200°C
Brand: Thermo Scientific Alfa Aesar A12728.22
100GR Silicone oil, for oil baths, usable rangefrom -40 to +200°C
Description
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10254541 | Certificate of Analysis | 25/03/2025 | A12728.22 |
10254542 | Certificate of Analysis | 25/03/2025 | A12728.22 |
10252956 | Certificate of Analysis | 04/11/2024 | A12728.36,A12728.22,A12728.0E |
10252957 | Certificate of Analysis | 04/11/2024 | A12728.36,A12728.22,A12728.0E |
10251930 | Certificate of Analysis | 06/08/2024 | A12728.36,A12728.22,A12728.0E |