Learn More
KGW Isotherm™ Dewar Cylindrical Insulated Flasks
For the storage and insulation of temperature-sensitive goods such as dry ice, liquid nitrogen and other coolants. KGW Isotherm™ Dewar Cylindrical Insulated Flasks feature a robust design made from borosilicate glass 3.3.
Brand:  KGW Isotherm™ 10215
 View more versions of this product
Product Code. 10169600
  • 2640.00 NOK / Each
Only null left
Estimated Shipment: 27-08-2025
Description
  • Easy handling
  • Cooling of small samples
  • Fully silvered as standard
  • Flexible applications in laboratory
  • Available with or without side grip
Specifications
Specifications
Blue
Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants
390mm
2 L
Borosilicate Glass
115mm
340mm
No side grip
90 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
KGW Isotherm™ Dewar Cylindrical Insulated Flasks > Capacity: 2000mL; Includes: No side grip; Inner Height: 340mm; Inner Diameter: 90mm