missing translation for 'onlineSavingsMsg'
Learn More

10-Undecen-1-ol, 99%, Thermo Scientific Chemicals

CAS: 112-43-6 | C11H22O | 170.296 g/mol

508.00 NOK - 6125.00 NOK

Chemical Identifiers

CAS 112-43-6
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00004750
InChI Key GIEMHYCMBGELGY-UHFFFAOYSA-N
Synonym 10-undecen-1-ol, undecylenic alcohol, undecylenyl alcohol, 10-undecenol, 1-undecen-11-ol, 10-undecylen-1-ol, 11-hydroxy-1-undecene, omega-undecenyl alcohol, undecen-1-ol, .omega.-undecenyl alcoholShow MoreShow Less
PubChem CID 8185
IUPAC Name undec-10-en-1-ol
SMILES C=CCCCCCCCCCO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11467856
SDS View Documents
Thermo Scientific Alfa Aesar
A14002.14
25 g
508.00 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
11477856
SDS View Documents
Thermo Scientific Alfa Aesar
A14002.22
100 g
1595.00 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
11487856
SDS View Documents
Thermo Scientific Alfa Aesar
A14002.36
500 g
6125.00 NOK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 203-971-0

missing translation for 'rtecsNumber' : YQ2985000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
10-Undecen-1-ol, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.