Learn More
BINDER™ 1 Year Warranty Extension
1 year warranty extension - The warranty is extended by 1 year from the delivery date, wear parts are excluded
7220.00 NOK - 18090.00 NOK
Specifications
Description | 1-year warranty extension - The warranty is extended by 1 year from the delivery date, wear parts are excluded |
---|---|
Duration | 1 Year |
Warranty and Service Offering | Extended Warranty |
Product Type | Extended Warranty (1 Year) |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
17079833
|
BINDER™
DL50-0010 |
BINDER Chambers Series B, BD, BD-S, BF, E, ED, ED-S, FD, FD-S, FED, FP, KB, KT, M |
7220.00 NOK
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
17089833
|
BINDER™
DL50-0020 |
BINDER Chambers Series VD, VDL |
9945.00 NOK
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
17099833
|
BINDER™
DL50-0030 |
BINDER Chambers Series KBW, KBWF, KBF, KBF P, KBF LQC, KMF, KBF-S |
14460.00 NOK
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
17009843
|
BINDER™
DL50-0040 |
BINDER Chambers Series MK, MKF, MKT, MKFT |
18090.00 NOK
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
Description
- Warranty extended by one year from delivery date
- Wear parts are excluded
Specifications
1-year warranty extension - The warranty is extended by 1 year from the delivery date, wear parts are excluded | |
1 Year | |
Extended Warranty | |
Extended Warranty (1 Year) |
Product Title
BINDER™ 1 Year Warranty Extension