Learn More
Masterflex™ Stopcocks with Luer Connections
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
499.00 NOK - 576.16 NOK
Specifications
Type | Stopcock |
---|---|
Material | Polycarbonate, HDPE |
Plug Material | Polycarbonate and HDPE |
Product Code | Brand | Connectors | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connectors | Price | Quantity & Availability | ||||||
11722683
|
Masterflex™
30600-00 |
Luer | N/A | |||||||
11772643
|
Masterflex™
30600-01 |
Male Luer Slip |
576.16 NOK
Pack of 10 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
Description
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
- Handle rotates 90°
- PC body and HDPE stem
Specifications
Stopcock | |
Polycarbonate, HDPE | |
Polycarbonate and HDPE |