missing translation for 'onlineSavingsMsg'
Learn More
1-Pyrrolidinecarbonyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11353027

1-Pyrrolidinecarbonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11353027
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11353027

Brand: Thermo Scientific Alfa Aesar L13396.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

Solubility
It reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-63-8
Molecular Formula C5H8ClNO
Molecular Weight (g/mol) 133.575
MDL Number MFCD00051321
InChI Key XACWJIQLDLUFSR-UHFFFAOYSA-N
Synonym 1-pyrrolidinecarbonyl chloride, pyrrolidinecarbonyl chloride, 1-pyrrolidinecarbonylchloride, pyrrolidine-1-carbonylchloride, pyrrolidinocarbonyl chloride, 1pyrrolidinecarbonyl chloride, pyrrolidine carbonyl chloride, acmc-1bo54, 1-pyrrolidine carbonylchloride, 1-pyrolidine carbonyl chloride
PubChem CID 70937
IUPAC Name pyrrolidine-1-carbonyl chloride
SMILES C1CCN(C1)C(=O)Cl

Specifications

Boiling Point 75°C (1mmHg)
Refractive Index 1.494
UN Number UN3265
Beilstein 110281
Solubility Information It reacts with water.
Density 1.209
Quantity 5 g
Flash Point 97°C (206°F)
Formula Weight 133.58
Sensitivity Moisture sensitive
Percent Purity 97%
Chemical Name or Material 1-Pyrrolidinecarbonyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10213077 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13396.03,L13396.06
Lot Number10193298 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13396.03,L13396.06
Lot NumberK16U019 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13396.03,L13396.06
Lot Number10228228 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13396.03,L13396.06
Lot NumberE29T041 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13396.03,L13396.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Pyrrolidinecarbonyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.