missing translation for 'onlineSavingsMsg'
Learn More
1-Propanesulfonyl chloride, 97% Product Code.: 15497238

1-Propanesulfonyl chloride, 97%

Product Code. p-7040862
25 mL, 25mL
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 15497238

Brand: Thermo Scientific Alfa Aesar H60747.AC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10147-36-1 | C3H7ClO2S | 142.597 g/mol

1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.

Solubility
Decomposes in hot water.

Notes
Moisture sensitive. Store in dry temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10147-36-1
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007462
InChI Key KPBSJEBFALFJTO-UHFFFAOYSA-N
Synonym 1-propanesulfonyl chloride, propanesulfonyl chloride, propanesulphonyl chloride, propylsulfonyl chloride, 1-propanesulfonylchloride, n-propanesulfonyl chloride, 1-propane sulfonyl chloride, chloropropylsulfone, n-prso2cl, propanesulfonylchloride
PubChem CID 66279
IUPAC Name propane-1-sulfonyl chloride
SMILES CCCS(=O)(=O)Cl

Specifications

Density 1.267
Boiling Point 78°C to 79°C
Flash Point 97°C (207°F)
Odor Pungent, Makes Eyes Water
Linear Formula CH3CH2CH2SO2Cl
Refractive Index 1.453
Quantity 25 mL
UN Number UN3265
Beilstein 1747500
Sensitivity Moisture sensitive
Solubility Information Decomposes in hot water.
Formula Weight 142.6
Percent Purity 97%
Chemical Name or Material 1-Propanesulfonyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberM18I060 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeH60747.AE,H60747.AC
Lot NumberT22G018 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeH60747.AE,H60747.AC
Lot NumberP22I022 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeH60747.AE,H60747.AC
Lot NumberS25E040 Certificate TypeCertificate of Analysis Date29/11/2021 Product CodeH60747.AE,H60747.AC
Lot NumberZ21E047 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH60747.AE,H60747.AC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Propanesulfonyl chloride, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.