Learn More
1-phenyl-1H-imidazole-2-carbaldehyde, Thermo Scientific™
Product Code. 10052214
1 g, Amber Glass Bottle, 1g
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
Chemical Identifiers
CAS 6002-15-9
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
InChI Key VJDRBJPZIHUDNE-UHFFFAOYSA-N
Synonym 1-phenyl-1h-imidazole-2-carbaldehyde, 1h-imidazole-2-carboxaldehyde, 1-phenyl, 1h-imidazole-2-carbaldehyde, 1-phenyl, 1-phenyl-2-formylimidazole, 1-phenylimidazole-2-carboxaldehyde
PubChem CID 6424701
IUPAC Name 1-phenylimidazole-2-carbaldehyde
SMILES C1=CC=C(C=C1)N2C=CN=C2C=O
Specifications
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 1-phenyl-1H-imidazole-2-carbaldehyde
Product Content Correction
Product Title
1-phenyl-1H-imidazole-2-carbaldehyde, Thermo Scientific™ >