missing translation for 'onlineSavingsMsg'
Learn More
1-Penten-3-ol, 97% Product Code.: 11450701

1-Penten-3-ol, 97%

Product Code. 11450701
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11450701

Brand: Thermo Scientific Alfa Aesar B20155.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 616-25-1 | C5H10O | 86.13 g/mol

1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-25-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00004573
InChI Key VHVMXWZXFBOANQ-UHFFFAOYNA-N
Synonym 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
PubChem CID 12020
IUPAC Name pent-1-en-3-ol
SMILES CCC(O)C=C

Specifications

Density 0.838
Boiling Point 114°C to 115°C
Flash Point 25°C (77°F)
Refractive Index 1.425
Quantity 5 g
UN Number UN1987
Beilstein 1719834
Solubility Information Slightly soluble in water.
Formula Weight 86.13
Percent Purity ≥98%
Chemical Name or Material 1-Penten-3-ol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Penten-3-ol, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.