missing translation for 'onlineSavingsMsg'
Learn More
1-Pentanesulfonic acid, sodium salt hydrate, 99% Product Code.: 10169322

1-Pentanesulfonic acid, sodium salt hydrate, 99%

Product Code. p-3737142 Shop All Thermo Scientific Chemicals Products
5 g, Glass bottle, 5g
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10169322

Brand: Thermo Scientific Acros 270370050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1266615-65-9 | C5H11NaO3S | 174.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1266615-65-9
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym n-Amylsulfonic acid, sodium salt
IUPAC Name sodium pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

Melting Point >300.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99%
Linear Formula CH3(CH2)4SO3Na·xH2O
Beilstein 04, III, 23
Solubility Information (10% in water) Clear colorless
Formula Weight 174.12
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 9.6% max. (100°C, 3 hrs) (vacuum dried)
Packaging Glass bottle
Appearance (10% aq. soln.) clear, colorless
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Pentanesulfonic acid, sodium salt hydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.