Learn More
1-Pentadecene, 97% Product Code.: 15410428
1-Pentadecene, 97%
Product Code. p-7035789
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15410428
Brand: Thermo Scientific Alfa Aesar H53516.06
Request Bulk or Custom Format
CAS: 13360-61-7 | C15H30 | 210.405 g/mol
1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13360-61-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00008987
InChI Key PJLHTVIBELQURV-UHFFFAOYSA-N
Synonym 1-pentadecene, pentadecene, pentadecene,1, unii-kh577lia74, ccris 5721, acmc-1bt6q
PubChem CID 25913
ChEBI CHEBI:77506
IUPAC Name pentadec-1-ene
SMILES CCCCCCCCCCCCCC=C
Specifications
Melting Point -4°C
Density 0.775
Boiling Point 268°C to 269°C
Flash Point 110°C (230°F)
Linear Formula CH3(CH2)12CH=CH2
Refractive Index 1.439
Quantity 5 g
Beilstein 1751011
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 210.4
Percent Purity 97%
Chemical Name or Material 1-Pentadecene
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberX13I018 Certificate TypeCertificate of Analysis Date28/10/2022 Product CodeH53516.06,H53516.14
Lot NumberP21I506 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeH53516.06,H53516.14
Lot Number5027R07W Certificate TypeCertificate of Analysis Date10/08/2020 Product CodeH53516.06,H53516.14
Lot NumberR26G041 Certificate TypeCertificate of Analysis Date10/08/2020 Product CodeH53516.06,H53516.14
Lot NumberM18E026 Certificate TypeCertificate of Analysis Date10/08/2020 Product CodeH53516.06,H53516.14
Product Title
1-Pentadecene, 97% >