missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-imidazole-5-carbohydrazide, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 140.15 |
Percent Purity | 97% |
Chemical Name or Material | 1-Methyl-1H-imidazole-5-carbohydrazide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'