missing translation for 'onlineSavingsMsg'
Learn More

1-Hexylpyridinium chloride, 97%, Thermo Scientific™

Product Code. 10564151 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle, 25g
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Ionic liquid
TRUSTED_SUSTAINABILITY

Specifications

CAS 6220-15-1
CAS Min % 96.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C11H18ClN
MDL Number MFCD03093294
Quantity 25 g
Synonym 1-hexylpyridinium chloride, 1-hexylpyridin-1-ium chloride, hexylpyridinium chloride, 1-hexylpyridiniumchloride, dsstox_cid_27911, dsstox_rid_82664, dsstox_gsid_47935
InChI Key JEOSMYVMLZTQOH-UHFFFAOYSA-M
SMILES CCCCCC[N+]1=CC=CC=C1.[Cl-]
IUPAC Name 1-hexylpyridin-1-ium;chloride
Molecular Weight (g/mol) 199.72
PubChem CID 2734172
Formula Weight 199.72
Percent Purity 97%
Physical Form Fused Solid
Chemical Name or Material 1-Hexylpyridinium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-Hexylpyridinium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Hexylpyridinium chloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.