missing translation for 'onlineSavingsMsg'
Learn More
1-Chloro-7-phenylheptane, 98+%, Thermo Scientific™ Product Code.: 11302736

1-Chloro-7-phenylheptane, 98+%, Thermo Scientific™

Product Code. 11302736
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11302736

Brand: Thermo Scientific Alfa Aesar L08817.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71434-47-4
Molecular Formula C13H19Cl
Molecular Weight (g/mol) 210.745
MDL Number MFCD00039392
InChI Key PNUBJDFTMKBZJK-UHFFFAOYSA-N
Synonym 1-chloro-7-phenylheptane, 7-chloroheptyl benzene, 7-phenyl-1-heptyl chloride, 7-phenylheptylchloride, 7-chloro-1-phenylheptane, 7-chloroheptyl benzene #, benzene, 7-chloroheptyl
PubChem CID 522410
IUPAC Name 7-chloroheptylbenzene
SMILES C1=CC=C(C=C1)CCCCCCCCl

Specifications

Boiling Point 94°C to 96°C (0.3 mmHg)
Refractive Index 1.505
Quantity 250 mg
Beilstein 2209292
Formula Weight 210.75
Percent Purity ≥98%
Chemical Name or Material 1-Chloro-7-phenylheptane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chloro-7-phenylheptane, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.