Learn More
1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals Product Code.: 11398966
1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals
Product Code. 11398966
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11398966
Brand: Thermo Scientific Alfa Aesar L20158.06
Request Bulk or Custom Format
It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60876-70-2
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.117
MDL Number MFCD00792676
InChI Key QIWQHUCUWNGYDZ-UHFFFAOYSA-N
Synonym 1-bromo-4-tert-butoxy benzene, 1-bromo-4-tert-butoxybenzene, 4-tert-butoxybromobenzene, 4-bromophenyl tert-butyl ether, 1-bromo-4-tert-butoxy-benzene, benzene, 1-bromo-4-1,1-dimethylethoxy, tert-butyl-4-bromophenyl ether, 1-bromo-4-2-methylpropan-2-yl oxy benzene, 1-tert-butoxy-4-bromobenzene, bromobutoxybenzene
PubChem CID 2763959
IUPAC Name 1-bromo-4-[(2-methylpropan-2-yl)oxy]benzene
SMILES CC(C)(C)OC1=CC=C(C=C1)Br
Specifications
Boiling Point 63°C to 65°C (0.4 mmHg)
Refractive Index 1.527
Quantity 5 g
Beilstein 2516411
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 229.12
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-tert-butoxybenzene
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number10243320 Certificate TypeCertificate of Analysis Date07/09/2022 Product CodeL20158.06,L20158.03,L20158.14
Lot Number10237452 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeL20158.06,L20158.03,L20158.14
Lot Number10224581 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeL20158.06,L20158.03,L20158.14
Lot Number10194934 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL20158.06,L20158.03,L20158.14
Lot Number10118397 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL20158.06,L20158.03,L20158.14
Product Title
1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals >