missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-3,5-di-tert-butylbenzene, 99% Product Code.: 11383626

1-Bromo-3,5-di-tert-butylbenzene, 99%

Product Code. 11383626
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11383626

Brand: Thermo Scientific Alfa Aesar L14646.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22385-77-9 | C14H21Br | 269.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22385-77-9
Molecular Formula C14H21Br
Molecular Weight (g/mol) 269.226
MDL Number MFCD00796945
InChI Key BUOWTUULDKULFI-UHFFFAOYSA-N
Synonym 1-bromo-3,5-di-tert-butylbenzene, 3,5-di-tert-butylbromobenzene, 3,5-di-t-butylbromobenzene, 1-bromo-3,5-di-t-butylbenzene, 1-bromo-3,5-ditert-butyl benzene, benzene, 1-bromo-3,5-bis 1,1-dimethylethyl, 1,5-bis tert-butyl-3-bromobenzene, acmc-1cpbo, g00018-watson-int, 3,5-di-t-butyl-bromobenzene
PubChem CID 620136
IUPAC Name 1-bromo-3,5-ditert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC(=C1)Br)C(C)(C)C

Specifications

Melting Point 62°C to 66°C
Quantity 1 g
Beilstein 2091559
Formula Weight 269.23
Percent Purity 99%
Chemical Name or Material 1-Bromo-3,5-di-tert-butylbenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10188515 Certificate TypeCertificate of Analysis Date10/07/2023 Product CodeL14646.06,L14646,L14646.03,L14646.14
Lot Number10187616 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeL14646.06,L14646.03,L14646.14
Lot Number10159505 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL14646.06,L14646.03,L14646.14
Lot Number10137571 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL14646.06,L14646.03,L14646.14
Lot Number10188515 Certificate TypeCertificate of Analysis Date06/12/2021 Product CodeL14646.06,L14646.03,L14646.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-3,5-di-tert-butylbenzene, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.