missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophen-3-ylmethylamine hydrochloride, 95%, Thermo Scientific™

3410.00 NOK - 3778.71 NOK

Chemical Identifiers

CAS 55810-74-7
Molecular Formula C9H10ClNS
Molecular Weight (g/mol) 199.696
MDL Number MFCD01566652
InChI Key MIAPJPCDRSPNPW-UHFFFAOYSA-N
Synonym benzo b thiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethylamine hydrochloride, benzo b thiophen-3-ylmethylamine hydrochloride, 1-1-benzothiophen-3-yl methanamine hydrochloride, benzo b thiophen-3-ylmethylamine, chloride, 1-benzothiophen-3-yl methanamine hydrochloride, 3-aminomethyl benzo b thiophene hydrochloride, benzo b thiophen-3-yl-methylamine hydrochloride salt, 1-1-benzothiophen-3-yl methanamine-hydrogen chloride 1/1
PubChem CID 2776346
IUPAC Name 1-benzothiophen-3-ylmethanamine;hydrochloride
SMILES C1=CC=C2C(=C1)C(=CS2)CN.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10723043
View Documents
Thermo Scientific Maybridge
CC12313DA
1 g
3410.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
11340567
View Documents
Thermo Scientific Maybridge
CC12313DE
5 g
3778.71 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

55810-74-7
199.696
MIAPJPCDRSPNPW-UHFFFAOYSA-N
2776346
C1=CC=C2C(=C1)C(=CS2)CN.Cl
C9H10ClNS
MFCD01566652
benzo b thiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethylamine hydrochloride, benzo b thiophen-3-ylmethylamine hydrochloride, 1-1-benzothiophen-3-yl methanamine hydrochloride, benzo b thiophen-3-ylmethylamine, chloride, 1-benzothiophen-3-yl methanamine hydrochloride, 3-aminomethyl benzo b thiophene hydrochloride, benzo b thiophen-3-yl-methylamine hydrochloride salt, 1-1-benzothiophen-3-yl methanamine-hydrogen chloride 1/1
1-benzothiophen-3-ylmethanamine;hydrochloride

Specifications

55810-74-7
100.0
Amber glass bottle
MFCD01566652
MIAPJPCDRSPNPW-UHFFFAOYSA-N
1-benzothiophen-3-ylmethanamine;hydrochloride
2776346
95%
80.0
95%
C9H10ClNS
benzo b thiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethylamine hydrochloride, benzo b thiophen-3-ylmethylamine hydrochloride, 1-1-benzothiophen-3-yl methanamine hydrochloride, benzo b thiophen-3-ylmethylamine, chloride, 1-benzothiophen-3-yl methanamine hydrochloride, 3-aminomethyl benzo b thiophene hydrochloride, benzo b thiophen-3-yl-methylamine hydrochloride salt, 1-1-benzothiophen-3-yl methanamine-hydrogen chloride 1/1
C1=CC=C2C(=C1)C(=CS2)CN.Cl
199.696
199.7
1-Benzothiophen-3-ylmethylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.