missing translation for 'onlineSavingsMsg'
Learn More
1,7,7-Trimethylbicyclo[2.2.1]heptan-2-amine hydrochloride, 97%, Thermo Scientific™ Product Code.: 10217762

1,7,7-Trimethylbicyclo[2.2.1]heptan-2-amine hydrochloride, 97%, Thermo Scientific™

Product Code. p-3484696
1 g, 1g
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10217762

Brand: Thermo Scientific Maybridge RDP00128DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 32768-19-7
Molecular Formula C10H20ClN
Molecular Weight (g/mol) 189.727
MDL Number MFCD00180693
InChI Key XVVITZVHMWAHIG-VMKQZJJLSA-N
Synonym 1,7,7-trimethylbicyclo 2.2.1 heptan-2-amine hydrochloride, bornylamine, hydrochloride, bornylamine hydrochloride, 1,7,7-trimethylnorbornan-2-amine, 1,7,7-trimethylbicyclo 2.2.1 hept-2-ylamine, chloride, 4,7,7-trimethylbicyclo 2.2.1 heptan-3-amine hydrochloride
PubChem CID 55283045
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-amine;hydrochloride
SMILES CC1(C2CCC1(C(C2)N)C)C.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 189.73
Percent Purity 97%
Chemical Name or Material 1, 7, 7-Trimethylbicyclo[2.2.1]heptan-2-amine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,7,7-Trimethylbicyclo[2.2.1]heptan-2-amine hydrochloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.